Please login to your online account to display your discounted pricing

R&D Systems Cotton Rat IL-6 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Molecular Weight 22 kDa
Name IL-6
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

561R6025

 
r&d systems
RDS561R6025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $428.87

561R6025CF

 
r&d systems
RDS561R6025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 22 kDa
Name IL-6
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Cotton Rat

Source: E. coli-derived, Leu25-Asn212, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Regulatory Status: RUO