Please login to your online account to display your discounted pricing

R&D Systems Cotton Rat IL-2 (Cys146Tyr) Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Molecular Weight 15.6 kDa
Name IL-2 (Cys146Tyr)
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 20μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

579R2020

 
r&d systems
RDS579R2020
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $419.81

579R2020CF

 
r&d systems
RDS579R2020CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 15.6 kDa
Name IL-2 (Cys146Tyr)
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 20μg
Recombinant Yes
Source Cotton Rat

Source: E. coli-derived, Ala21-Gln155 (Cys146Tyr), with and without an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Regulatory Status: RUO