missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Cotton Rat IL-13 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Molecular Weight 13.1 kDa
Name IL-13
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
6940CL025 R&D Systems
RDS6940CL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
6940CL025CF R&D Systems
RDS6940CL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Met1-His133, with a C-terminal 5-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IL-13
25μg
Cotton Rat
13.1 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes