Please login to your online account to display your discounted pricing

R&D Systems Cotton Rat IL-10 (Cys149Tyr) Recombinant Protein

$389.76 - $389.76

Specifications

Conjugate Unlabeled
Molecular Weight 18.7 kDa (monomer)
Name IL-10 (Cys149Tyr)
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

581CR010

 
r&d systems
RDS581CR010
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose and with BSA as a carrier protein. Each for $389.76

581CR010CF

 
r&d systems
RDS581CR010CF
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. Each for $389.76
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 18.7 kDa (monomer)
Name IL-10 (Cys149Tyr)
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Cotton Rat

Source: E. coli-derived, Ser1-Ser160 (Cys149Tyr)
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.7 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Regulatory Status: RUO