missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Cotton Rat IL-1 beta/IL-1F2 Recombinant Protein

$337.62 - $337.62

Specifications

Conjugate Unlabeled
Molecular Weight 17.3 kDa
Name IL-1 beta/IL-1F2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1009CL010 R&D Systems
RDS1009CL010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $337.62
1009CL010CF R&D Systems
RDS1009CL010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $337.62
Specifications

Description

Source: E. coli-derived, Val116-Ser267, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IL-1 beta/IL-1F2
10μg
Cotton Rat
17.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes