missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Cotton Rat IL-1 alpha/IL-1F1 Recombinant Protein

$328.11 - $328.11

Specifications

Conjugate Unlabeled
Molecular Weight 17.7 kDa
Name IL-1 alpha/IL-1F1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
552R1010CF R&D Systems
RDS552R1010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $328.11
552R1010 R&D Systems
RDS552R1010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $328.11
Specifications

Description

Source: E. coli-derived, Ser115-Pro269
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IL-1 alpha/IL-1F1
10μg
Cotton Rat
17.7 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes