Please login to your online account to display your discounted pricing

R&D Systems Cotton Rat IFN-gamma Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Molecular Weight 17 kDa
Name IFN-gamma
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

565RG025

 
r&d systems
RDS565RG025
Lyophilized from a 0.2μm filtered solution in PBS, NaCl and Sucrose with BSA as a carrier protein. Each for $376.73

565RG025CF

 
r&d systems
RDS565RG025CF
Supplied as a 0.2μm filtered solution in PBS, NaCl and Sucrose. Each for $376.73
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 17 kDa
Name IFN-gamma
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Cotton Rat

Source: E. coli-derived, His21-Ile170, with an N-terminal Met and two substitutions, G150R, Y165N
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Supplied as a 0.2μm filtered solution in PBS, NaCl and Sucrose.
Regulatory Status: RUO