missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Cotton Rat IFN-alpha (Cys109Tyr) Recombinant Protein

$366.12 - $366.12

Specifications

Conjugate Unlabeled
Molecular Weight 19.4 kDa
Name IFN-alpha (Cys109Tyr)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1082CR025 R&D Systems
RDS1082CR025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $366.12
1082CR025CF R&D Systems
RDS1082CR025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $366.12
Specifications

Description

Source: E. coli-derived, Cys24-Lys190 (Cys109Tyr)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IFN-alpha (Cys109Tyr)
25μg
Cotton Rat
19.4 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes