Please login to your online account to display your discounted pricing

R&D Systems Cotton Rat IFN-alpha (Cys109Tyr) Recombinant Protein


Manufacturer: r&d systems  RDS1082CR025CF

 View more versions of this product

Catalog No. 1082CR025CF

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 19.4 kDa
Name IFN-alpha (Cys109Tyr)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Cotton Rat
Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.