Please login to your online account to display your discounted pricing

R&D Systems Cotton Rat CXCL2/GRO beta/MIP-2/CINC-3 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Molecular Weight 8.0 kDa
Name CXCL2/GRO beta/MIP-2/CINC-3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1084M2025

 
r&d systems
RDS1084M2025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87

1084M2025CF

 
r&d systems
RDS1084M2025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 8.0 kDa
Name CXCL2/GRO beta/MIP-2/CINC-3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Cotton Rat

Source: E. coli-derived, Ala29-Asn101
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.0 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO