missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Chicken Netrin-2 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 396387
Molecular Weight 64 kDa
Name Netrin-2
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
127N2025CF R&D Systems
RDS127N2025CF
Lyophilized from a 0.2μm filtered solution in PBS and NaCl. Each for $416.79
127N2025 R&D Systems
RDS127N2025
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ala16-Pro581, with a C-terminal 6-His tag & Ala16-Pro581, with an N-terminal Met and C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 64 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Entrez Gene ID: 396387
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
64 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
396387
Netrin-2
25μg
Chicken