missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Canine VEGF Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 403802
Molecular Weight 19 kDa (monomer)
Name VEGF
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1603CV010 R&D Systems
RDS1603CV010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $428.87
1603CV010CF R&D Systems
RDS1603CV010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $428.87
Specifications

Description

Source: E. coli-derived, Pro28-Arg190
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 403802
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
19 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
403802
VEGF
10μg
Canine