Please login to your online account to display your discounted pricing

R&D Systems Canine KGF/FGF-7 Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 403915
Molecular Weight 18.8 kDa
Name KGF/FGF-7
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1957KG025

 
r&d systems
RDS1957KG025
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT with BSA as a carrier protein. Each for $419.81

1957KG025CF

 
r&d systems
RDS1957KG025CF
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 403915
Molecular Weight 18.8 kDa
Name KGF/FGF-7
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Canine

Source: E. coli-derived, Cys32-Thr194 & Ser55-Thr194
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT.
Entrez Gene ID: 403915
Regulatory Status: RUO