missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Canine IL-6 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 403985
Molecular Weight 20.8 kDa
Name IL-6
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1609CL025 R&D Systems
RDS1609CL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
1609CL025CF R&D Systems
RDS1609CL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: E. coli-derived, Thr23-Met207, with an N-terminal Met & Pro24-Met207
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 403985
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
20.8 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
403985
IL-6
25μg
Canine