Please login to your online account to display your discounted pricing

R&D Systems Canine IL-4 Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 403785
Molecular Weight 12.9 kDa
Name IL-4
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

754CL025

 
r&d systems
RDS754CL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $419.81

754CL025CF

 
r&d systems
RDS754CL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 403785
Molecular Weight 12.9 kDa
Name IL-4
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Canine

Source: E. coli-derived, His25-His132, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 403785
Regulatory Status: RUO