missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Canine IL-17/IL-17A Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 481837
Molecular Weight 15.2 kDa (monomer)
Name IL-17/IL-17A
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
5848CL025 R&D Systems
RDS5848CL025
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. Each for $428.87
5848CL025CF R&D Systems
RDS5848CL025CF
Lyophilized from a 0.2μm filtered solution in HCl. Each for $428.87
Specifications

Description

Source: E. coli-derived, Gly81-Ala210
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.2 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in HCl.
Entrez Gene ID: 481837
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15.2 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
481837
IL-17/IL-17A
25μg
Canine