missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Canine IL-12 Recombinant Protein

$467.98 - $467.98

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 403977
Molecular Weight 36.3 kDa (p40), 23.6 kDa (p35)
Name IL-12
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2118CL025 R&D Systems
RDS2118CL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $467.98
2118CL025CF R&D Systems
RDS2118CL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $467.98
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Ile23-Ser329 (p40) & Arg26-Ser222 (p35), both wih a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36.3 kDa (p40), 23.6 kDa (p35)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 403977
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
36.3 kDa (p40), 23.6 kDa (p35)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
403977
IL-12
25μg
Canine