missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Canine IL-10 Recombinant Protein

$389.76 - $389.76

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 403628
Molecular Weight 18.9 kDa
Name IL-10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
735CL010 R&D Systems
RDS735CL010
Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl with BSA as a carrier protein. Each for $389.76
735CL010CF R&D Systems
RDS735CL010CF
Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl. Each for $389.76
Specifications

Description

Source: E. coli-derived, Ser20-Ile179
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl.
Entrez Gene ID: 403628
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
403628
IL-10
10μg
Canine