missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Canine IFN-gamma Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 403801
Molecular Weight 17 kDa
Name IFN-gamma
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
781CG050CF R&D Systems
RDS781CG050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $376.73
781CG050 R&D Systems
RDS781CG050
Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol and Tween™ 80 with BSA as a carrier protein. Each for $376.73
Specifications

Description

Source: E. coli-derived, Gln24-Lys166, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 403801
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
403801
IFN-gamma
50μg
Canine