missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Canine Erythropoietin Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Molecular Weight 18.4 kDa
Name Erythropoietin
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3816CE010 R&D Systems
RDS3816CE010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
3816CE010CF R&D Systems
RDS3816CE010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ala41-Arg206
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
Erythropoietin
10μg
Canine
18.4 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes