missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Canine CXCL8/IL-8 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 403850
Molecular Weight 9 kDa (mature), 8.5 kDa (N-terminal truncation)
Name CXCL8/IL-8
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1608CL025 R&D Systems
RDS1608CL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
1608CL025CF R&D Systems
RDS1608CL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: E. coli-derived, Ala23-Pro101 & Val28-Pro101
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9 kDa (mature), 8.5 kDa (N-terminal truncation)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 403850
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
9 kDa (mature), 8.5 kDa (N-terminal truncation)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
403850
CXCL8/IL-8
25μg
Canine