Please login to your online account to display your discounted pricing

R&D Systems Canine CCL2/JE/MCP-1 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 403981
Molecular Weight 8.8 kDa
Name CCL2/JE/MCP-1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1774MC025CF

 
r&d systems
RDS1774MC025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87

1774MC025

 
r&d systems
RDS1774MC025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 403981
Molecular Weight 8.8 kDa
Name CCL2/JE/MCP-1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Canine

Source: E. coli-derived, Gln24-Pro101
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 403981
Regulatory Status: RUO