missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Bovine TNF-alpha Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 280943
Molecular Weight 17.6 kDa
Name TNF-alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2279BT025CF R&D Systems
RDS2279BT025CF
Supplied as a 0.2μm filtered solution in PBS. Each for $376.73
2279BT025 R&D Systems
RDS2279BT025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $376.73
Specifications

Description

Source: E. coli-derived, Leu78-Leu234, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.6 kDa
Formulation: Supplied as a 0.2μm filtered solution in PBS.
Entrez Gene ID: 280943
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
280943
TNF-alpha
25μg
Bovine