missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Bovine Fibronectin Protein

$181.19 - $428.87

Specifications

Conjugate Unlabeled
Formulation Supplied as a 0.2μm filtered solution in Tris-HCl, NaCl and Urea.
Gene ID (Entrez) 280794
Name Fibronectin
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
1030FN05M R&D Systems
RDS1030FN05M
5mg Each for $428.87
1030FN01M R&D Systems
RDS1030FN01M
1mg Each for $181.19
Specifications

Description

Source: Bovine plasma-derived
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Supplied as a 0.2μm filtered solution in Tris-HCl, NaCl and Urea.
Entrez Gene ID: 280794
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
280794
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Supplied as a 0.2μm filtered solution in Tris-HCl, NaCl and Urea.
Fibronectin
Bovine