missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Bovine FGF basic Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 281161
Molecular Weight 16.5 kDa
Name FGF basic
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2099FB025 R&D Systems
RDS2099FB025
Lyophilized from a 0.2μm filtered solution in Tris-HCl with BSA as a carrier protein. Each for $298.51
2099FB025CF R&D Systems
RDS2099FB025CF
Lyophilized from a 0.2μm filtered solution in Tris-HCl and trehalose. Each for $298.51
Specifications

Description

Source: E. coli-derived, Pro10-Ser155, with an N-terminal Ala
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Tris-HCl andtrehalose.
Entrez Gene ID: 281161
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
16.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
281161
FGF basic
25μg
Bovine