missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Bovine FGF basic Protein

$520.12 - $520.12

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 281161
Name FGF basic
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
133FB025CF R&D Systems
RDS133FB025CF
Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl. Each for $520.12
133FB025 R&D Systems
RDS133FB025
Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein. Each for $520.12
Specifications

Description

Source: Bovine brain tissue-derived
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa, reducing conditions
Formulation: Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
Entrez Gene ID: 281161
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
FGF basic
25μg
281161
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Bovine