missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Bovine FGF acidic Protein

$467.98 - $467.98

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 281160
Name FGF acidic
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
132FA025 R&D Systems
RDS132FA025
Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein. Each for $467.98
132FA025CF R&D Systems
RDS132FA025CF
Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl. Each for $467.98
Specifications

Description

Source: Bovine brain tissue-derived
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl.
Entrez Gene ID: 281160
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
FGF acidic
25μg
281160
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Bovine