Please login to your online account to display your discounted pricing
R&D Systems b-AP15/VLX1500
Specifically inhibits the 19S regulatory particle-associated deubiquitinases USP14 and UCHL5, resulting in the accumulation of intracellular polyubiquitin
Manufacturer: r&d systems  I19701M
 View more versions of this product
Catalog No. I19701M
  • / Each
Description & Specifications
Specifications
Molecular Weight 419.39
Quantity 1mg
Product Type 19S proteasome inhibitor
Inhibitors 19S proteasome