Please login to your online account to display your discounted pricing

R&D Systems b-AP15/VLX1500

Specifically inhibits the 19S regulatory particle-associated deubiquitinases USP14 and UCHL5, resulting in the accumulation of intracellular polyubiquitin

$71.69 - $71.69

Specifications

Inhibitors 19S proteasome
Molecular Weight 419.39
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

I19701M

 
r&d systems
I19701M
1mg Each for $71.69
Description & Specifications

Specifications

Inhibitors 19S proteasome
Molecular Weight 419.39

b-AP15 has been shown to inhibit tumor progression in at least four solid tumor models, and also inhibits organ infiltration in an acute myeloid leukemia (AML) model.

  • Potent, reversible, cell-permeable inhibitor
  • Purity >97% by HPLC
  • Decreases HCT-116 cell viability at 72 hours (EC50 = 0.4μM)