Please login to your online account to display your discounted pricing

(R)-CPP, Tocris


Potent NMDA antagonist; More active enantiomer of (RS)-CPP

Quantity 10mg

Manufacturer: r & d systems  024710

 View more versions of this product

Catalog No. 024710

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 252.21
Molecular Formula C8H17N2O5P
Assay Percent Range >98%
CAS 126453-07-4
Recommended Storage Desiccate at Room Temperature
Synonym 3-((R)-2-Carboxypiperazin-4-yl)-propyl-1-phosphonic acid
Chemical Name or Material (R)-CPP

Specifications

Chemical Name or Material (R)-CPP
Assay Percent Range >98%
Molecular Formula C8H17N2O5P
Formula Weight 252.21
Synonym 3-((R)-2-Carboxypiperazin-4-yl)-propyl-1-phosphonic acid
Accessories

Desiccate at Room Temperature