Please login to your online account to display your discounted pricing

(R)-CPP, Tocris

Potent NMDA antagonist; More active enantiomer of (RS)-CPP

$447.38 - $1,831.50

Chemical Identifiers

Chemical Name or Material (R)-CPP
Assay Percent Range >98%
Molecular Formula C8H17N2O5P
Formula Weight 252.21
Synonym 3-((R)-2-Carboxypiperazin-4-yl)-propyl-1-phosphonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

024710

 
r & d systems
024710
10mg Each for $447.38

024750

 
r & d systems
024750
50mg Each for $1,831.50
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 252.21
Chemical Name or Material (R)-CPP
Assay Percent Range >98%
Molecular Formula C8H17N2O5P
CAS 126453-07-4
Synonym 3-((R)-2-Carboxypiperazin-4-yl)-propyl-1-phosphonic acid
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature