Please login to your online account to display your discounted pricing

(R)-BOC-2,4-Dichlorophenylalanine, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC368830050

 View more versions of this product

Catalog No. AC368830050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Powder
Formula Weight 334.2
Quantity 5g
Color White to off-white
Specific Rotation Condition + 32.00 (20.00°C c=1,methanol)
Packaging Glass bottle
Chemical Name or Material (R)-BOC-2, 4-Dichlorophenylalanine
Specific Rotation +30° to +34° (20°C, 589nm) (C=1, methanol)
Molecular Formula C14 H17 Cl2 N O4
MDL Number MFCD01862941
CAS 114873-12-0
Synonym N-tert-Butoxycarbonyl-2, 4-dichloro-D-phenylalanine
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (R)-BOC-2, 4-Dichlorophenylalanine
Assay ≥98.5%
Molecular Formula C14 H17 Cl2 N O4
Formula Weight 334.2
MDL Number MFCD01862941
Synonym N-tert-Butoxycarbonyl-2, 4-dichloro-D-phenylalanine
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents