Please login to your online account to display your discounted pricing

(R)-BOC-2,4-Dichlorophenylalanine, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC368830010

 View more versions of this product

Catalog No. AC368830010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Powder
Formula Weight 334.2
Quantity 1g
Color White to off-white
Specific Rotation Condition + 32.00 (20.00°C c=1,methanol)
Infrared Spectrum Authentic
Specific Rotation +30° to +34° (20°C, 589nm) (C=1, methanol)
MDL Number MFCD01862941
Packaging Glass bottle
Synonym N-tert-Butoxycarbonyl-2, 4-dichloro-D-phenylalanine
Chemical Name or Material (R)-BOC-2, 4-Dichlorophenylalanine
CAS 114873-12-0
Molecular Formula C14 H17 Cl2 N O4
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

Specifications

Chemical Name or Material (R)-BOC-2, 4-Dichlorophenylalanine
Assay ≥98.5%
Molecular Formula C14 H17 Cl2 N O4
Formula Weight 334.2
MDL Number MFCD01862941
Synonym N-tert-Butoxycarbonyl-2, 4-dichloro-D-phenylalanine
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents