Please login to your online account to display your discounted pricing

(R)-BOC-2,4-Dichlorophenylalanine, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-BOC-2, 4-Dichlorophenylalanine
Assay ≥98.5%
Molecular Formula C14 H17 Cl2 N O4
Formula Weight 334.2
MDL Number MFCD01862941
Synonym N-tert-Butoxycarbonyl-2, 4-dichloro-D-phenylalanine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC368830050

  SDS
thermo fisher scientific
AC368830050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Powder
Formula Weight 334.2
Quantity 5g
Color White to off-white
Specific Rotation Condition + 32.00 (20.00°C c=1,methanol)
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C14 H17 Cl2 N O4
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD01862941
Specific Rotation +30° to +34° (20°C, 589nm) (C=1, methanol)
Synonym N-tert-Butoxycarbonyl-2, 4-dichloro-D-phenylalanine
Chemical Name or Material (R)-BOC-2, 4-Dichlorophenylalanine
CAS 114873-12-0