missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(+)-Bis-(1,2-diphenylphosphino)propane, 93%, Tech., ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 254902500

 View more versions of this product

Catalog No. AC254902500



Specifications

Specifications

(R)-(+)-Bis-(1, 2-diphenylphosphino)propane
Glass Bottle
White
250mg
67884-32-6
100.0
C27H26P2
MFCD00012309
WGOBPPNNYVSJTE-HSZRJFAPSA-N
[(2R)-1-diphenylphosphanylpropan-2-yl]-diphenylphosphane
2724539
Granular Powder
Technical
Authentic
+170° to +175° (20°C, 589nm) (c=1, acetone)
68°C to 73°C
tech., 93%
91.0
93%
(C6H5)2PCH(CH3)CH2P(C6H5)2
r-+-1,2-bis diphenylphosphino propane, r-+-1,2bis diphenylphosphino propane, r-+-bis 1,2-diphenylphosphino propane, r-+-bis-1,2-diphenylphosphino propane, r-+-prophos, r-+-propylenebis 1,2-diphenylphosphine, r-propane-1,2-diylbis diphenylphosphine, r-prophos
CC(CP(C1=CC=CC=C1)C2=CC=CC=C2)P(C3=CC=CC=C3)C4=CC=CC=C4
412.453
412.45
≥91%

Chemical Identifiers

67884-32-6
412.453
WGOBPPNNYVSJTE-HSZRJFAPSA-N
2724539
CC(CP(C1=CC=CC=C1)C2=CC=CC=C2)P(C3=CC=CC=C3)C4=CC=CC=C4
C27H26P2
MFCD00012309
r-+-1,2-bis diphenylphosphino propane, r-+-1,2bis diphenylphosphino propane, r-+-bis 1,2-diphenylphosphino propane, r-+-bis-1,2-diphenylphosphino propane, r-+-prophos, r-+-propylenebis 1,2-diphenylphosphine, r-propane-1,2-diylbis diphenylphosphine, r-prophos
[(2R)-1-diphenylphosphanylpropan-2-yl]-diphenylphosphane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

AIR SENSITIVE

TSCA : TSCA

SDS Documents