Learn More
(R)-(+)-Bis-(1,2-diphenylphosphino)propane, 93%, Tech., ACROS Organics™
$93.86 - $93.86
Chemical Identifiers
CAS | 67884-32-6 |
---|---|
Molecular Formula | C27H26P2 |
Molecular Weight (g/mol) | 412.45 |
MDL Number | MFCD00012309 |
InChI Key | WGOBPPNNYVSJTE-HSZRJFAPSA-N |
Synonym | r-+-1,2-bis diphenylphosphino propane, r-+-1,2bis diphenylphosphino propane, r-+-bis 1,2-diphenylphosphino propane, r-+-bis-1,2-diphenylphosphino propane, r-+-prophos, r-+-propylenebis 1,2-diphenylphosphine, r-propane-1,2-diylbis diphenylphosphine, r-prophos |
PubChem CID | 2724539 |
IUPAC Name | [(2R)-1-diphenylphosphanylpropan-2-yl]-diphenylphosphane |
SMILES | CC(CP(C1=CC=CC=C1)C2=CC=CC=C2)P(C3=CC=CC=C3)C4=CC=CC=C4 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC254902500
|
Thermo Fisher Scientific
254902500 |
250mg | Glass Bottle |
Each for $93.86
|
|
AC254900010
|
Thermo Fisher Scientific
254900010 |
1g | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
67884-32-6 | |
412.45 | |
WGOBPPNNYVSJTE-HSZRJFAPSA-N | |
2724539 | |
CC(CP(C1=CC=CC=C1)C2=CC=CC=C2)P(C3=CC=CC=C3)C4=CC=CC=C4 |
C27H26P2 | |
MFCD00012309 | |
r-+-1,2-bis diphenylphosphino propane, r-+-1,2bis diphenylphosphino propane, r-+-bis 1,2-diphenylphosphino propane, r-+-bis-1,2-diphenylphosphino propane, r-+-prophos, r-+-propylenebis 1,2-diphenylphosphine, r-propane-1,2-diylbis diphenylphosphine, r-prophos | |
[(2R)-1-diphenylphosphanylpropan-2-yl]-diphenylphosphane |
Specifications
Authentic | |
+170° to +175° (20°C, 589nm) (c=1, acetone) | |
68°C to 73°C | |
67884-32-6 | |
100.0 | |
C27H26P2 | |
MFCD00012309 | |
WGOBPPNNYVSJTE-HSZRJFAPSA-N | |
[(2R)-1-diphenylphosphanylpropan-2-yl]-diphenylphosphane | |
2724539 | |
Granular Powder | |
Technical |
Glass Bottle | |
White | |
250mg | |
91.0 | |
93% | |
(C6H5)2PCH(CH3)CH2P(C6H5)2 | |
r-+-1,2-bis diphenylphosphino propane, r-+-1,2bis diphenylphosphino propane, r-+-bis 1,2-diphenylphosphino propane, r-+-bis-1,2-diphenylphosphino propane, r-+-prophos, r-+-propylenebis 1,2-diphenylphosphine, r-propane-1,2-diylbis diphenylphosphine, r-prophos | |
CC(CP(C1=CC=CC=C1)C2=CC=CC=C2)P(C3=CC=CC=C3)C4=CC=CC=C4 | |
412.453 | |
412.45 | |
≥91% | |
(R)-(+)-Bis-(1, 2-diphenylphosphino)propane, tech., 93% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : TSCA