Please login to your online account to display your discounted pricing

(R)-(+)-Bay K 8644, Tocris

Ca2+-channel blocker (L-type)

$323.08 - $1,333.44

Chemical Identifiers

Chemical Name or Material (R)-(+)-Bay K 8644
Assay Percent Range >97%
Molecular Formula C16H15F3N2O4
Formula Weight 356.3
Synonym (4R)-1, 4-Dihydro-2, 6-dimethyl-5-nitro-4-[2-trifluoromethyl)phenyl]-3-pyridinecarboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

154510

 
r & d systems
RDS154510
10mg Each for $323.08

154550

 
r & d systems
RDS154550
50mg Each for $1,333.44
Description & Specifications

Specifications

Formula Weight 356.3
Quantity 10mg
Recommended Storage Store at +4°C
CAS 98791-67-4
Synonym (4R)-1, 4-Dihydro-2, 6-dimethyl-5-nitro-4-[2-trifluoromethyl)phenyl]-3-pyridinecarboxylic acid methyl ester
Molecular Formula C16H15F3N2O4
Assay Percent Range >97%
Chemical Name or Material (R)-(+)-Bay K 8644
Safety and Handling

Store at +4°C