Please login to your online account to display your discounted pricing

(R)-Baclofen, Tocris

Selective GABAB agonist; Active enantiomer of (RS)-Baclofen

$116.01 - $451.04

Chemical Identifiers

Chemical Name or Material (R)-Baclofen
Assay Percent Range >99%
Molecular Formula C10H12ClNO2
Formula Weight 213.66
Synonym (R)-4-Amino-3-(4-chlorophenyl)butanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

079610

 
r & d systems
RDS079610
10mg Each for $116.01

079650

 
r & d systems
RDS079650
50mg Each for $451.04
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 213.66
Chemical Name or Material (R)-Baclofen
Assay Percent Range >99%
Molecular Formula C10H12ClNO2
CAS 69308-37-8
Synonym (R)-4-Amino-3-(4-chlorophenyl)butanoic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature