missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(+)-alpha-Amino-gamma-butyrolactone hydrochloride monohydrate, 97%, ACROS Organics™

Chemical Identifiers

CAS 104347-13-9
Molecular Formula C4H7NO2·ClH
Molecular Weight (g/mol) 155.59
InChI Key XBKCXPRYTLOQKS-AENDTGMFSA-N
Synonym 2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r, 3r-3-aminotetrahydrofuran-2-one hydrochloride, a-amino-, a-butyrolactone hydrochloride, d-homoserine lactone hcl, d-homoserine lactone hydrochloride, homoserine lactone hydrochloride, pubchem14743, r-+-, r-+-a-amino-g-butyrolactone hydrochloride, r-+-alpha-amino-gamma-butyrolactone hydrochloride, r-3-aminodihydrofuran-2 3h-one hydrochloride
PubChem CID 11954435
IUPAC Name (3R)-3-aminooxolan-2-one;hydrochloride
SMILES C1COC(=O)C1N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC437570010
SDS View Documents
Thermo Fisher Scientific
AC437570010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC437570050
SDS View Documents
Thermo Fisher Scientific
AC437570050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

104347-13-9
155.59
2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r, 3r-3-aminotetrahydrofuran-2-one hydrochloride, a-amino-, a-butyrolactone hydrochloride, d-homoserine lactone hcl, d-homoserine lactone hydrochloride, homoserine lactone hydrochloride, pubchem14743, r-+-, r-+-a-amino-g-butyrolactone hydrochloride, r-+-alpha-amino-gamma-butyrolactone hydrochloride, r-3-aminodihydrofuran-2 3h-one hydrochloride
(3R)-3-aminooxolan-2-one;hydrochloride
C4H7NO2·ClH
XBKCXPRYTLOQKS-AENDTGMFSA-N
11954435
C1COC(=O)C1N.Cl

Specifications

29.64 to 32.12% C, 8.64 to 9.36% N
Glass bottle
15% max.
215.1°C to 217.1°C
104347-13-9
100.0
C4H7NO2·ClH
XBKCXPRYTLOQKS-AENDTGMFSA-N
(3R)-3-aminooxolan-2-one;hydrochloride
11954435
Powder, Solid or Crystalline Powder
(R)-(+)-α-Amino-γ-butyrolactone hydrochloride monohydrate
Authentic
30
Yellow to Brown
1g
96.0
96.0 to max. 104% min. (Argentometry)
2 3h-furanone, 3-aminodihydro-, hydrochloride, 3r, 3r-3-aminotetrahydrofuran-2-one hydrochloride, a-amino-, a-butyrolactone hydrochloride, d-homoserine lactone hcl, d-homoserine lactone hydrochloride, homoserine lactone hydrochloride, pubchem14743, r-+-, r-+-a-amino-g-butyrolactone hydrochloride, r-+-alpha-amino-gamma-butyrolactone hydrochloride, r-3-aminodihydrofuran-2 3h-one hydrochloride
C1COC(=O)C1N.Cl
155.59
155.59
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Warning