Please login to your online account to display your discounted pricing

(R)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic, 99%(e.e.>99.5%), ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC312220010

 View more versions of this product

Catalog No. AC312220010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Low Melting Solid
Formula Weight 234.17
Quantity 1g
Color White
Infrared Spectrum Authentic
Specific Rotation +69° to +73° (20°C, 589nm) (c=1.6, CH3OH)
Name Note 99%(e.e.>99.5%)
Molecular Formula C10H9F3O3
MDL Number MFCD00004184
CAS 20445-31-2
Synonym (+)MTPA
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Refrigerator +4°C
Chemical Name or Material (R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
Linear Formula C6H5C(OCH3)(CF3)CO2H
Packaging Glass bottle

Specifications

Chemical Name or Material (R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
Name Note 99%(e.e.>99.5%)
CAS 20445-31-2
Assay ≥98.5%
Molecular Formula C10H9F3O3
Linear Formula C6H5C(OCH3)(CF3)CO2H
Formula Weight 234.17
MDL Number MFCD00004184
Synonym (+)MTPA
Accessories

Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

Refrigerator +4°C

SDS & Safety Documents