(R)-α-Allylalanine hydrate, 98%, 98% ee, ACROS Organics™
Manufacturer:  Acros Organics AC441255000
 View more versions of this product
Catalog No. AC441255000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
(R)-α-Allylalanine hydrate
Authentic
White to Yellow
98% ee
97.5
98%
(R)-(+)-2-Amino-2-methyl-4-pentenoic acid hydrate
Powder
98.0% min.
+24° to +28° (20°C, 589nm) (c=0.88 in water)
500mg
1266371-17-8,96886-56-5
100.0
C6H11NO2·xH2O
129.16
97.5 to 102.5% as monohydrate
Chemical Identifiers
1266371-17-8
C6H11NO2·xH2O
(R)-(+)-2-Amino-2-methyl-4-pentenoic acid hydrate
SDS Documents