Please login to your online account to display your discounted pricing

(R)-α-Allylalanine hydrate, 98%, 98% ee, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-α-Allylalanine hydrate
Name Note 98% ee
CAS 1266371-17-8
CAS Min % 97.5
CAS Max % 100.0
CAS 96886-56-5
Assay 97.5 to 102.5% as monohydrate
Assay Percent Range 98%
Molecular Formula C6H11NO2·xH2O
Formula Weight 129.16
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC441251000

 
thermo fisher scientific
AC441251000
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC441255000

 
thermo fisher scientific
AC441255000
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 129.16
Quantity 100mg
Color White to pale yellow
Assay 97.5 to 102.5% as monohydrate
CAS Max % 100.0
Name Note 98% ee
CAS 1266371-17-8
Specific Rotation +24° to +28° (20°C, 589nm) (c=0.88 in water)
Chemical Name or Material (R)-α-Allylalanine hydrate
Assay Percent Range 98%
Molecular Formula C6H11NO2·xH2O
CAS Min % 97.5
CAS 96886-56-5
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Synonym (R)-(+)-2-Amino-2-methyl-4-pentenoic acid hydrate