Please login to your online account to display your discounted pricing

R-96544 hydrochloride, Tocris

Potent, selective 5-HT2A antagonist

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material R-96544 hydrochloride
Assay Percent Range >99%
Molecular Formula C22H29NO3·HCl
Formula Weight 391.94
Synonym (2R, 4R)-5-[2-[2-[2-(3-Methoxyphenyl)ethyl]phenoxy]ethyl]-1-methyl-3-pyrrolidinol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

174250

 
r & d systems
174250
50mg Each for $915.10

174210

 
r & d systems
174210
10mg Each for $218.74
Description & Specifications

Specifications

Formula Weight 391.94
Quantity 50mg
Chemical Name or Material R-96544 hydrochloride
Assay Percent Range >99%
Molecular Formula C22H29NO3·HCl
CAS 167144-80-1
Synonym (2R, 4R)-5-[2-[2-[2-(3-Methoxyphenyl)ethyl]phenoxy]ethyl]-1-methyl-3-pyrrolidinol hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature