Please login to your online account to display your discounted pricing

(R)-(+)-8-Hydroxy-DPAT hydrobromide, Tocris


Selective 5-HT1A agonist; Enantiomer of 8-Hydroxy-DPAT hydrobromide

Quantity 10mg

Manufacturer: r & d systems  RDS108010

 View more versions of this product

Catalog No. 108010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 328.29
Quantity 10mg
Chemical Name or Material (R)-(+)-8-Hydroxy-DPAT hydrobromide
Molecular Formula C16H25NO·HBr
CAS 78095-19-9
Assay Percent Range >99%
Synonym (2R)-(+)-8-Hydroxy-2-(di-n-propylamino)tetralin hydrobromide
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material (R)-(+)-8-Hydroxy-DPAT hydrobromide
Assay Percent Range >99%
Molecular Formula C16H25NO·HBr
Formula Weight 328.29
Synonym (2R)-(+)-8-Hydroxy-2-(di-n-propylamino)tetralin hydrobromide
Accessories

Desiccate at +4°C