Please login to your online account to display your discounted pricing

(R)-(+)-8-Hydroxy-DPAT hydrobromide, Tocris

Selective 5-HT1A agonist; Enantiomer of 8-Hydroxy-DPAT hydrobromide

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material (R)-(+)-8-Hydroxy-DPAT hydrobromide
Assay Percent Range >99%
Molecular Formula C16H25NO·HBr
Formula Weight 328.29
Synonym (2R)-(+)-8-Hydroxy-2-(di-n-propylamino)tetralin hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

108050

 
r & d systems
RDS108050
50mg Each for $862.95

108010

 
r & d systems
RDS108010
10mg Each for $205.70
Description & Specifications

Specifications

Formula Weight 328.29
Quantity 50mg
Chemical Name or Material (R)-(+)-8-Hydroxy-DPAT hydrobromide
Assay Percent Range >99%
Molecular Formula C16H25NO·HBr
CAS 78095-19-9
Synonym (2R)-(+)-8-Hydroxy-2-(di-n-propylamino)tetralin hydrobromide
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C