Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-6-Methyl-5-hepten-2-ol, 99%

$69.46 - $201.91

Chemical Identifiers

Chemical Name or Material (R)-(-)-6-Methyl-5-hepten-2-ol
CAS 58917-27-4
Molecular Formula C8H16O
Formula Weight 128.22
MDL Number MFCD03093079
Beilstein 1720072
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1888303

  SDS
alfa aesar™
L1888303
1g Each for $201.91

AAL18883MD

  SDS
alfa aesar™
L18883MD
250mg Each for $69.46
Description & Specifications

Specifications

Formula Weight 128.22
Chemical Name or Material (R)-(-)-6-Methyl-5-hepten-2-ol
Assay 99%
MDL Number MFCD03093079
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Molecular Formula C8H16O
CAS 58917-27-4
Optical Rotation −18° (Neat)
Health Hazard 1 Warning
Quantity 1g
Boiling Point 78° to 80°C (15mmHg)
Flash Point 67°C (152°F)
Beilstein 1720072
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.