Please login to your online account to display your discounted pricing

(R)-5-Phthalimido-2-bromovaleric acid, 95% (95% e.e.), ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC365700010

 View more versions of this product

Catalog No. AC365700010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 326.15
Physical Form Powder
Quantity 1g
Color Off-white
Enantiomeric Excess 94% min.
Name Note 95%, (95% e.e.)
CAS 179090-36-9
Chemical Name or Material (R)-5-Phthalimido-2-bromovaleric acid
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C13H12BrNO4
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
MDL Number MFCD03427676

Specifications

Chemical Name or Material (R)-5-Phthalimido-2-bromovaleric acid
Name Note 95%, (95% e.e.)
CAS 179090-36-9
Assay ≥94%
Molecular Formula C13H12BrNO4
Formula Weight 326.15
MDL Number MFCD03427676
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents