missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-5-Phthalimido-2-bromovaleric acid, 95%,(95% ee), Acros Organics

Chemical Identifiers

CAS 179090-36-9
Molecular Formula C13H12BrNO4
Molecular Weight (g/mol) 326.15
MDL Number MFCD03427676
InChI Key FCGBPUWRRBLODE-SNVBAGLBSA-N
Synonym 2r-2-bromo-5-1,3-dioxoisoindol-2-yl pentanoic acid
PubChem CID 2734540
IUPAC Name (2R)-2-bromo-5-(1,3-dioxoisoindol-2-yl)pentanoic acid
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCC(C(=O)O)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC365700010 SDS View Documents Thermo Fisher Scientific
AC365700010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

179090-36-9
326.15
FCGBPUWRRBLODE-SNVBAGLBSA-N
2734540
C1=CC=C2C(=C1)C(=O)N(C2=O)CCCC(C(=O)O)Br
C13H12BrNO4
MFCD03427676
2r-2-bromo-5-1,3-dioxoisoindol-2-yl pentanoic acid
(2R)-2-bromo-5-(1,3-dioxoisoindol-2-yl)pentanoic acid

Specifications

94% min.
Glass bottle
1g
C13H12BrNO4
2r-2-bromo-5-1,3-dioxoisoindol-2-yl pentanoic acid
C1=CC=C2C(=C1)C(=O)N(C2=O)CCCC(C(=O)O)Br
326.15
326.15
95%
Authentic
White
179090-36-9
MFCD03427676
FCGBPUWRRBLODE-SNVBAGLBSA-N
(2R)-2-bromo-5-(1,3-dioxoisoindol-2-yl)pentanoic acid
2734540
Powder
(R)-5-Phthalimido-2-bromovaleric acid, 95%, (95% e.e.)
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning