missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-5-(Hydroxymethyl)-2-pyrrolidinone 98.0+%, TCI America™

$148.13 - $518.45

Chemical Identifiers

CAS 66673-40-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00077791
InChI Key HOBJEFOCIRXQKH-SCSAIBSYSA-N
Synonym --d-pyroglutamol, 5-hydroxymethyl-2-pyrrolidinone, 5r-5-hydroxymethyl pyrrolidin-2-one, d-pyroglutaminol, r---5-hydroxymethyl-2-pyrrolidinone, r---5-hydroxymethylpyrrolidin-2-one, r-5-hydroxymethyl pyrrolidin-2-one, r-5-hydroxymethyl-2-pyrrolidinone, r-5-hydroxymethyl-pyrrolidin-2-one
PubChem CID 7128291
IUPAC Name (5R)-5-(hydroxymethyl)pyrrolidin-2-one
SMILES C1CC(=O)NC1CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H12411G View Documents Tci America
H12411G
1g Each for $148.13
 
H12415G View Documents Tci America
H12415G
5g Each for $518.45
 
Specifications

Chemical Identifiers

66673-40-3
115.132
HOBJEFOCIRXQKH-SCSAIBSYSA-N
7128291
C1CC(=O)NC1CO
C5H9NO2
MFCD00077791
--d-pyroglutamol, 5-hydroxymethyl-2-pyrrolidinone, 5r-5-hydroxymethyl pyrrolidin-2-one, d-pyroglutaminol, r---5-hydroxymethyl-2-pyrrolidinone, r---5-hydroxymethylpyrrolidin-2-one, r-5-hydroxymethyl pyrrolidin-2-one, r-5-hydroxymethyl-2-pyrrolidinone, r-5-hydroxymethyl-pyrrolidin-2-one
(5R)-5-(hydroxymethyl)pyrrolidin-2-one

Specifications

White-Yellow
1g
C5H9NO2
--d-pyroglutamol, 5-hydroxymethyl-2-pyrrolidinone, 5r-5-hydroxymethyl pyrrolidin-2-one, d-pyroglutaminol, r---5-hydroxymethyl-2-pyrrolidinone, r---5-hydroxymethylpyrrolidin-2-one, r-5-hydroxymethyl pyrrolidin-2-one, r-5-hydroxymethyl-2-pyrrolidinone, r-5-hydroxymethyl-pyrrolidin-2-one
C1CC(=O)NC1CO
115.132
115.13
≥98.0% (GC,N)
84°C
66673-40-3
MFCD00077791
HOBJEFOCIRXQKH-SCSAIBSYSA-N
(5R)-5-(hydroxymethyl)pyrrolidin-2-one
7128291
Crystal-Powder at 20°C
(R)-5-(Hydroxymethyl)-2-pyrrolidinone
Safety and Handling

Safety and Handling

TSCA : No