Please login to your online account to display your discounted pricing

(R)-(+)-[(5,6),(5',6')-bis(ethylenedioxy)biphenyl-2,2'-diyl](diphenylphosphine), 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-(+)-[(5,6),(5';,6';)-Bis(ethylenedioxy)biphenyl-2,2';-diyl](diphenylphosphine)
CAS 445467-61-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C40H32O4P2
Formula Weight 638.64
MDL Number MFCD05861608
Synonym (R)-Synphos™
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC375142500

  SDS
thermo fisher scientific
AC375142500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 638.64
Quantity 250mg
Color White
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C40H32O4P2
CAS Min % 97.5
Chemical Name or Material (R)-(+)-[(5,6),(5';,6';)-Bis(ethylenedioxy)biphenyl-2,2';-diyl](diphenylphosphine)
Packaging Glass bottle
MDL Number MFCD05861608
Synonym (R)-Synphos™
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 445467-61-8